Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) chloride hexahydrate, MilliporeSigma™

Catalog No. M1039430250 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1039430250 250 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1039430250 Supplier MilliporeSigma Supplier No. 1.03943.0250
Add to Cart
Add to Cart

For analysis EMSURE™ ACS, Reag. Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate; Iron trichloride
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Quantity 250 g
Solubility Information Soluble in water: 920g/L at 10°C
Formula Weight 270.33
Odor Chlorine
Packaging Plastic Bottle
Chemical Name or Material Iron(III) chloride hexahydrate
Hazard Category H302: Harmful if swallowed.
Hazard Statement H315: Causes skin irritation.
Precautionary Statement H318: Causes serious eye damage.
EINECSNumber 231-729-4
RTECSNumber NO5425000
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.