Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(III) Chloride Solution

Catalog No. 71085750ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-085-750ML 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-085-750ML Supplier Sigma Aldrich Supplier No. 71085750ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. FeCl3 is used in many biomass conversion processes because of its unique properties such as nontoxicity, low-cost, abundance, and high selectivity. Here, this product is 0.2 M in 2-methyltetrahydrofuran and thus aligns with Safer Solvents and Auxiliaries, Use of Renewable Feedstocks, and has been enhanced for catalytic efficiency. Click here for more information.Iron(III) chloride in greener solvent, 2-methyltetrahydrofuran (2-MeTHF) can be used: As a mild oxidant for phenolic coupling reaction. In dimerizing aryllithiumsand ketone enolates.As a mild Lewis acid catalyst to catalyzes ene reactions.In Nazarov cyclizations, Michael additions, and acetonation reactions.2-Methyltetrahydrofuran (2-MeTHF): A Biomass-Derived Solvent with Broad Application in Organic Chemistry

Specifications

CAS 173035-11-5
Melting Point 79°C to 85°C
Molecular Formula C21H26N2
Refractive Index n20/D 1.414
Linear Formula C21H26N2
MDL Number MFCD11973794
Quantity 250 mg
Synonym Ferric chloride solution
Molecular Weight (g/mol) 306.44 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.