Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) citrate tribasic monohydrate 18-20 Fe basis (T), 18-20% Fe basis (T), MilliporeSigma™ Supelco™

Catalog No. 111013620 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3620 1 kg
11-101-3618 250 g
11-101-3619 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-3620 Supplier MilliporeSigma Supelco Supplier No. 449411KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00013098
Synonym Ferric citrate monohydrate

Specifications

CAS 2338-05-8
Quantity 1 kg
Linear Formula C6H5 FeO7 · H2O
Formula Weight 262.96
Percent Purity 18 to 20% Fe basis (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.