Learn More
Medchemexpress LLC Iron(III) ferrocyanide; Milori blue | 14038-43-8 | 99.0% | 859.23 | C18Fe7N18 | 25 G
SDP
Catalog No. 5000240059
Encompass_Preferred
Prussian blue insoluble (Iron(III) ferrocyanide) is an inorganic pigment and chelating agent supplied as a fine, insoluble powder for research applications. It is commonly used as an adsorbent in studies of radiocesium and thallium decorporation and is provided with supporting quality documentation and analytical data, including CAS 14038-43-8, molecular formula C18Fe7N18, molecular weight 859.23, and reported purity ≥99.0%.
  • High purity (≥99.0%).
  • Insoluble powder form suitable for adsorption studies.
  • Documented quality with datasheet, COA, and SDS available.
  • Multiple package sizes available for laboratory needs.
  • Suitable as a pigment and chelating agent in remediation research.
Catalog No. 50-002-40059 Supplier Medchemexpress LLC Supplier No. HY106594A25G
Prussian blue insoluble (Iron(III) ferrocyanide) is an inorganic pigment and chelating agent supplied as a fine, insoluble powder for research applications. It is commonly used as an adsorbent in studies of radiocesium and thallium decorporation and is provided with supporting quality documentation and analytical data, including CAS 14038-43-8, molecular formula C18Fe7N18, molecular weight 859.23, and reported purity ≥99.0%.
  • High purity (≥99.0%).
  • Insoluble powder form suitable for adsorption studies.
  • Documented quality with datasheet, COA, and SDS available.
  • Multiple package sizes available for laboratory needs.
  • Suitable as a pigment and chelating agent in remediation research.