Learn More
Strem, An Ascensus Company CAS# 7783-50-8. 50g. Iron(III) fluoride, anhydrous, 99+%. MFCD00011012
SDP
Catalog No. 5090114635
Encompass
CAS# 7783-50-8. 50g. Iron(III) fluoride, anhydrous, 99+%. MFCD00011012. Molecular Weight: 112.84. Molecular Formula: FeF3. Color/form: greenish-brown pwdr. Strem# 93-2610. http://www.strem.com/catalog/v/93-2610/
Catalog No. 50-901-14635 Supplier Strem, An Ascensus Company Supplier No. 93261050G
CAS# 7783-50-8. 50g. Iron(III) fluoride, anhydrous, 99+%. MFCD00011012. Molecular Weight: 112.84. Molecular Formula: FeF3. Color/form: greenish-brown pwdr. Strem# 93-2610. http://www.strem.com/catalog/v/93-2610/