Learn More
Iron(III) fluoride, anhydrous, 97% min
Description
Iron(III) fluroide is used in ceramics manufacturing. It is used as a catalyst for certain cross coupling reactions. Further, it is involved in the chemoselective addition of cyanide to aldehyde to prepare cyanohydrins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | >1000° (sublimation) |
| Color | Green |
| Physical Form | Powder |
| Quantity | 50 g |
| Assay Percent Range | 97% min. (Metal basis) |
| Solubility Information | Soluble in dilute hydrofluroic acid. Slightly soluble in water. Insoluble in alcohol,ether and benzene. |
| Formula Weight | 112.84 |
| Sensitivity | Hygroscopic |
| Appearance | Pale green powder |
| Density | 3.87 |
| Show More |
Safety and Handling
| Hazard Category | H301-H311-H332-H314-H318: Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage. |
| EINECSNumber | 232-002-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.