Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iron(III) meso-tetra(4-carboxyphenyl)porphine chloride | 55266-17-6 | MFCD03425968 | 95.0% | 880.06 g·mol⁻¹ | C48H28ClFeN4O8 | 250 MG
SDP

Catalog No. 5000209667
Encompass_Preferred

FeTCPP chloride is an iron(III) porphyrin complex used as a catalyst and reagent in catalytic, electrochemical, photochemical, and biomedical research. It demonstrates strong visible-light photocatalytic activity and exhibits peroxidase-like (bionic catalysis) activity.

  • High purity (95.0%) suitable for research applications.
  • Molecular weight 880.06 g·mol⁻¹ and formula C48H28ClFeN4O8.
  • Available in laboratory quantities (250 mg, 500 mg, 1 g).
  • Effective visible-light photocatalyst for pollutant degradation studies.
  • Shows peroxidase-like catalytic activity for bionic catalysis research.

Catalog No. 50-002-09667 Supplier Medchemexpress LLC Supplier No. 250MGHYW075707
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

FeTCPP chloride is an iron(III) porphyrin complex used as a catalyst and reagent in catalytic, electrochemical, photochemical, and biomedical research. It demonstrates strong visible-light photocatalytic activity and exhibits peroxidase-like (bionic catalysis) activity.

  • High purity (95.0%) suitable for research applications.
  • Molecular weight 880.06 g·mol⁻¹ and formula C48H28ClFeN4O8.
  • Available in laboratory quantities (250 mg, 500 mg, 1 g).
  • Effective visible-light photocatalyst for pollutant degradation studies.
  • Shows peroxidase-like catalytic activity for bionic catalysis research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.