Learn More
Strem, An Ascensus Company CAS# 16456-81-8. 500mg. Iron(III) meso-tetraphenylporphine chloride. MFCD00012152
SDP
Catalog No. 5090111125
Encompass
CAS# 16456-81-8. 500mg. Iron(III) meso-tetraphenylporphine chloride. MFCD00012152. Molecular Weight: 704.03. Molecular Formula: Fe(C44H28N4)Cl. Color/form: purple xtl. Strem# 26-2880. http://www.strem.com/catalog/v/26-2880/
Catalog No. 50-901-11125 Supplier Strem, An Ascensus Company Supplier No. 262880500MG
CAS# 16456-81-8. 500mg. Iron(III) meso-tetraphenylporphine chloride. MFCD00012152. Molecular Weight: 704.03. Molecular Formula: Fe(C44H28N4)Cl. Color/form: purple xtl. Strem# 26-2880. http://www.strem.com/catalog/v/26-2880/