Iron(III) nitrate nonahydrate, ACS, 98.0-101.0%, Alfa Aesar™

Manufacturer: Alfa Aesar 33315F7

Quantity 5 x 2kg

 View more versions of this product

Catalog No. AA33315F7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7782-61-8
    Physical Form Crystalline
    Assay Percent Range 98.0 to 101.0%
    Molecular Formula FeN3O9·9H2O
    Formula Weight 403.99 (241.86 Anhydrous)
    MDL Number MFCD00149708
    Chemical Name or Material Iron(III) nitrate nonahydrate, For ACS analysis
    Grade ACS Grade

    Specifications

    Formula Weight 403.99 (241.86 Anhydrous)
    Quantity 5 x 2kg
    Color Pale purple
    DOT Information DOT Class: 5.1, Packing Group: III
    EINECS Number 233-899-5
    Physical Form Crystalline
    Assay Percent Range 98.0 to 101.0%
    Appearance Pale purple
    MDL Number MFCD00149708
    CAS 7782-61-8
    Odor Acidic
    Molecular Formula FeN3O9·9H2O
    Health Hazard 1 H272-H315-H319: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation.
    Chemical Name or Material Iron(III) nitrate nonahydrate, For ACS analysis
    Grade ACS Grade

    H272-H315-H319: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit