Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(Iii) Nitrate Nonahydrate, Solstice

Catalog No. 6000852 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-008-52 500 g
60-008-48 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-008-52 Supplier Solstice Supplier No. 216828500G

missing translation for 'validMainframeMsg'

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+);trinitrate;nonahydrate

Specifications

Quantity 500 g
Linear Formula Fe(NO3)3 · 9H2O
UN Number 3260
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Formula Weight 404g/mol
Percent Purity ≥98%
Grade ACS Reagent
Chemical Name or Material Iron(Iii) Nitrate Nonahydrate
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314
EINECSNumber 233-899-5
RTECSNumber NO7175000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.