Learn More
Medchemexpress LLC Iron(III) octaethylporphine chloride | 28755-93-3 | 95.0% | 624.06 g/mol | C36H44ClFeN4 | 5 MG
SDP
Catalog No. 5000283020
Encompass_Preferred
Iron(III) octaethylporphine chloride is an iron-centered metalloporphyrin supplied as a solid reagent for coordination chemistry, catalysis, and spectroscopic studies. Chemical formula C36H44ClFeN4, molecular weight 624.06 g/mol, CAS 28755-93-3. Supplied in small milligram quantities for laboratory research use only.
  • Metalloporphyrin model compound for heme-like studies.
  • High purity (95.0%) suitable for research applications.
  • Available in small milligram pack sizes for method development.
  • Solid form, stable under sealed, refrigerated storage.
  • Iron(III) center with chloride ligand enables coordination chemistry experiments.
Catalog No. 50-002-83020 Supplier Medchemexpress LLC Supplier No. HYW1278135MG
Iron(III) octaethylporphine chloride is an iron-centered metalloporphyrin supplied as a solid reagent for coordination chemistry, catalysis, and spectroscopic studies. Chemical formula C36H44ClFeN4, molecular weight 624.06 g/mol, CAS 28755-93-3. Supplied in small milligram quantities for laboratory research use only.
  • Metalloporphyrin model compound for heme-like studies.
  • High purity (95.0%) suitable for research applications.
  • Available in small milligram pack sizes for method development.
  • Solid form, stable under sealed, refrigerated storage.
  • Iron(III) center with chloride ligand enables coordination chemistry experiments.