Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iron(III) octaethylporphine chloride | 28755-93-3 | MFCD00011614 | 95.0% | 624.06 | C36H44ClFeN4 | 25 MG
SDP

Catalog No. 5000283290
Encompass_Preferred

Iron(III) octaethylporphine chloride is a metalloporphyrin reagent used in biochemical and life-science research. It is the chloro complex of octaethylporphyrin with iron(III), supplied as a solid with a listed purity of 95.0% and documented identification data including CAS 28755-93-3, molecular formula C36H44ClFeN4, and molecular weight 624.06.

  • Used as a metalloporphyrin reagent in biochemical studies.
  • Purity 95.0% suitable for research applications.
  • Molecular formula C36H44ClFeN4 and molecular weight 624.06 for compound identification.
  • Available in small milligram pack sizes for laboratory experiments.
  • Store sealed at 4°C; in solvent, store at -80°C for long-term storage.

Catalog No. 50-002-83290 Supplier Medchemexpress LLC Supplier No. HYW12781325MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iron(III) octaethylporphine chloride is a metalloporphyrin reagent used in biochemical and life-science research. It is the chloro complex of octaethylporphyrin with iron(III), supplied as a solid with a listed purity of 95.0% and documented identification data including CAS 28755-93-3, molecular formula C36H44ClFeN4, and molecular weight 624.06.

  • Used as a metalloporphyrin reagent in biochemical studies.
  • Purity 95.0% suitable for research applications.
  • Molecular formula C36H44ClFeN4 and molecular weight 624.06 for compound identification.
  • Available in small milligram pack sizes for laboratory experiments.
  • Store sealed at 4°C; in solvent, store at -80°C for long-term storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.