Learn More
Medchemexpress LLC Iron(III) octaethylporphine chloride | 28755-93-3 | MFCD00011614 | 95.0% | 624.06 g·mol⁻¹ | C36H44ClFeN4 | 10 MG
SDP
Catalog No. 5000282625
Encompass_Preferred
Iron(III) octaethylporphine chloride is a porphyrin coordination complex used as a biochemical reagent and model compound for heme-like systems. It is commonly applied in spectroscopic investigations, catalysis research, and mechanistic studies of iron porphyrin chemistry.
  • Used as a porphyrin model compound for heme-like systems.
  • Suitable for spectroscopic and mechanistic studies.
  • Typical purity ~95.0%.
  • Brown to black solid, easy to handle and store.
  • Provided in a 10 mg quantity for small-scale research.
Catalog No. 50-002-82625 Supplier Medchemexpress LLC Supplier No. HYW12781310MG
Iron(III) octaethylporphine chloride is a porphyrin coordination complex used as a biochemical reagent and model compound for heme-like systems. It is commonly applied in spectroscopic investigations, catalysis research, and mechanistic studies of iron porphyrin chemistry.
  • Used as a porphyrin model compound for heme-like systems.
  • Suitable for spectroscopic and mechanistic studies.
  • Typical purity ~95.0%.
  • Brown to black solid, easy to handle and store.
  • Provided in a 10 mg quantity for small-scale research.