Learn More
Medchemexpress LLC Iron(III) octaethylporphine chloride | 28755-93-3 | MFCD00011614 | 95.0% | 624.06 g/mol | C36H44ClFeN4 | 100 MG
SDP
Catalog No. 5000283677
Encompass_Preferred
Iron(III) octaethylporphine chloride is a metalloporphyrin reagent comprising an iron(III) center coordinated to an octaethylporphyrin ligand with a chloride axial ligand. It is supplied as a solid and is commonly used in catalysis research, heme-model studies, and spectroscopic characterization of porphyrin complexes.
  • Metalloporphyrin complex with a defined iron(III) oxidation state.
  • Suitable for catalysis, redox, and oxygen activation studies.
  • Useful as a heme model for biochemical and mechanistic research.
  • Provided as a solid with high purity suitable for research use.
  • Characterized by chemical formula and CAS number for material tracking.
Catalog No. 50-002-83677 Supplier Medchemexpress LLC Supplier No. HYW127813100MG
Iron(III) octaethylporphine chloride is a metalloporphyrin reagent comprising an iron(III) center coordinated to an octaethylporphyrin ligand with a chloride axial ligand. It is supplied as a solid and is commonly used in catalysis research, heme-model studies, and spectroscopic characterization of porphyrin complexes.
  • Metalloporphyrin complex with a defined iron(III) oxidation state.
  • Suitable for catalysis, redox, and oxygen activation studies.
  • Useful as a heme model for biochemical and mechanistic research.
  • Provided as a solid with high purity suitable for research use.
  • Characterized by chemical formula and CAS number for material tracking.