Learn More
Strem, An Ascensus Company CAS# 19469-07-9. 25g. Iron(III) oxalate hexahydrate, tech. gr.. MFCD00151243
SDP
Catalog No. 5090114642
Encompass
CAS# 19469-07-9. 25g. Iron(III) oxalate hexahydrate, tech. gr.. MFCD00151243. Molecular Weight: 375.76 (483.86). Molecular Formula: Fe2(C2O4)3∙6H2O. Color/form: yellow pwdr. Strem# 93-2615. http://www.strem.com/catalog/v/93-2615/
Catalog No. 50-901-14642 Supplier Strem, An Ascensus Company Supplier No. 93261525G
CAS# 19469-07-9. 25g. Iron(III) oxalate hexahydrate, tech. gr.. MFCD00151243. Molecular Weight: 375.76 (483.86). Molecular Formula: Fe2(C2O4)3∙6H2O. Color/form: yellow pwdr. Strem# 93-2615. http://www.strem.com/catalog/v/93-2615/