Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(III) oxide

Catalog No. 5293115G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-931-15G 5 g
52-931-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 52-931-15G Supplier Sigma Aldrich Supplier No. 5293115G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron is present in the form of iron(III) oxide in almost all natural sources of water. It is the most common supplement of iron for algae. It is a highly stable n-type semiconductor, and possesses a band gap of 2.0 to 2.2eV. It is widely used in solar photoelectrolysis cells. Self-organized nanoporous iron(III) oxide (haematite) has been prepared. Orientation effects on nearly opaque single crystals of iron(III) oxide have been investigated by laser Raman spectroscopy.Iron(III) oxide may be used in the preparation of iron(III) oxide-hydrate gels.

Specifications

CAS 1309-37-1
Quantity 5 g
Molecular Formula Fe2O3
Linear Formula Fe2O3
MDL Number MFCD00011008
Synonym Ferric oxide
Molecular Weight (g/mol) 159.69
Percent Purity ≥99.995% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.