Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) oxide, 99.85+% (metals basis), Thermo Scientific Chemicals

Numéro de catalogue. AA3638730
Modifier l'affichage
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AA3638730 250 g
AA3638718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AA3638730 Fournisseur Thermo Scientific Chemicals Code fournisseur 03638730
Ajouter au panier
Ajouter au panier
Request Bulk or Custom Formats

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

It has many applications, mainly used in iron industry, pigment, as photocatalyst, medicine and polishing jewels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Spécifications

Melting Point 1565°C
Color Red to Brown
Physical Form Sintered Lump
Quantity 250 g
Assay Percent Range >99.85% (Metals basis)
Solubility Information Insoluble in water,Soluble in diluted acids,sugar solution and slightly soluble in aq. tartaric acid,citric acid,CH3COOH.
Formula Weight 159.69
Odor Odorless
Density 5.24
Chemical Name or Material Iron(III) oxide
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber 215-168-2
RTECSNumber NO7400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.