Learn More
Strem, An Ascensus Company CAS# 1309-37-1. 50g. Iron(III) oxide (99.995%-Fe) PURATREM. MFCD00011008
SDP
Catalog No. 5090111115
Encompass
CAS# 1309-37-1. 50g. Iron(III) oxide (99.995%-Fe) PURATREM. MFCD00011008. Molecular Weight: 159.69. Molecular Formula: Fe2O3. Color/form: red to brown pwdr. Strem# 26-2750. http://www.strem.com/catalog/v/26-2750/
Catalog No. 50-901-11115 Supplier Strem, An Ascensus Company Supplier No. 26275050G
CAS# 1309-37-1. 50g. Iron(III) oxide (99.995%-Fe) PURATREM. MFCD00011008. Molecular Weight: 159.69. Molecular Formula: Fe2O3. Color/form: red to brown pwdr. Strem# 26-2750. http://www.strem.com/catalog/v/26-2750/