Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(Iii) Oxide, purified, Solstice

Catalog No. 6001927 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-27 1 kg
60-019-28 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-019-27 Supplier Solstice Supplier No. 123421KG
Only null left
Add to Cart
Add to Cart

Purified,≥95%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-37-1
Molecular Formula Fe2O3
Molecular Weight (g/mol) 159.69
MDL Number MFCD00011008
InChI Key JEIPFZHSYJVQDO-UHFFFAOYSA-N
IUPAC Name trioxodiiron
SMILES [Fe][Fe](=O)(=O)=O

Specifications

Quantity 1 kg
UN Number NONH for all modes of transport
Formula Weight 159.69g/mol
Percent Purity ≥95%
Grade Purified
Chemical Name or Material Iron(Iii) Oxide
EINECSNumber 215-168-2
RTECSNumber NO7400000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.