Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(III) P-Toluenesulfonate Hexahydrate

Catalog No. 46286125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
46-286-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 46-286-125G Supplier Sigma Aldrich Supplier No. 46286125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron(III) p-toluenesulfonate hexahydrate (Fe(CH3C6H4SO3)3-+6H2O) can be used:As an oxidant in the synthesis of poly(3,4-ethylenedioxythiophene) (PEDOT) nanofilms by vapor phase polymerization.In the synthesis of iron(III) complex of pyridoxal-4-methylthiosemicarbazone.As a precursor material for the synthesis of Fe, N, and S functionalized carbon electrocatalyst for the oxygen reduction reaction.As an oxidant in the synthesis of poly(thiophene phenylenes) (PThP).

Specifications

CAS 4336-70-3
Melting Point 275°C
Molecular Formula C20H17ClNP
Linear Formula NCCH2P(C6H5)3Cl
Quantity 25 g
MDL Number MFCD00031672
Molecular Weight (g/mol) 337.78 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.