Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) phosphate hydrate

Catalog No. AA31106A1
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA31106A1 1 kg
AA3110630 250 g
AA31106A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA31106A1 Supplier Thermo Scientific Chemicals Supplier No. 031106A1
Only null left
Add to Cart

CAS: 51833-68-2 | FeH2O5P | 168.83 g/mol

Poly(methyl methacrylate) is used in the lenses of exterior lights of automobile and automotive components such as badges, steering wheel, tachometer covers, warning triangles and fascia panels. It is in the form of sheets used in shatter resistant panels for building windows, skylights, bullet proof security barriers, bathtubs, LCD screens, furniture and many other applications. It is used as the matrix in laser dye-doped organic solid-state gain media for tunable solid state dye lasers. Further, it is used as a shield to stop beta radiation emitted from radioisotopes, as dosimeter devices during the gamma irradiation process and as a dispersant for ceramic powders to stabilize colloidal suspensions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51833-68-2
Molecular Formula FeH2O5P
Molecular Weight (g/mol) 168.83
MDL Number MFCD00016096
InChI Key RQKNQWUJROKKEQ-UHFFFAOYSA-K
Synonym ferric phosphate hydrate, iron iii phosphate hydrate, ferric orthophosphate hydrate, ccris 8095, acmc-20akpj, fe.po4.h2o, ksc497s7j, iron 3+ hydrate phosphate, iron 3+ ion hydrate phosphate, phosphoric acid, iron 3 salt 1:1 , hydrate
PubChem CID 21410239
SMILES O.[Fe+3].[O-]P([O-])([O-])=O

Specifications

Color White
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water.
IUPAC Name iron(3+);phosphate;hydrate
Formula Weight 150.82 (anhydrous)
Appearance Off-white powder
Chemical Name or Material Iron(III) phosphate hydrate, hydrate
Hazard Category H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
EINECSNumber 233-149-7
RTECSNumber TC6190000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.