Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron(III) pyrophosphate

Catalog No. 503125596 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5596 100 g
50-312-6083 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-5596 Supplier Sigma Aldrich Supplier No. P6526100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ferric Phosphate is a tan or yellowish white odorless powder. It is insoluble in water and soluble in mineral acids. It can be prepared by reacting sodium pyrophosphate with ferric citrate. It shows good bioaccesibility.

Specifications

CAS 10058-44-3
Quantity 100 g
Molecular Formula Fe4O21P6
Linear Formula Fe4(P2O7)3
MDL Number MFCD00016091
Synonym Diphosphoric acid iron(III) salt; Ferric pyrophosphate
Molecular Weight (g/mol) 745.21
Percent Purity 10.5-12.5% (titration)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.