Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron(III) sulfate hydrate, Puriss. p.a., 21-23% Fe basis, Solstice

Catalog No. 6000857 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 1 kg
Packaging:
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-008-57 500 g Poly bottle
60-045-925 1 kg Poly bottle
60-045-926 6 x 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-008-57 Supplier Solstice Supplier No. 31235500G
Add to Cart
Add to Cart

Puriss. p.a., 21-23% Fe basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-10-7
Molecular Formula Fe2H2O13S3
Molecular Weight (g/mol) 417.87
MDL Number MFCD00011007
InChI Key VXWSFRMTBJZULV-UHFFFAOYSA-H
PubChem CID 167265
IUPAC Name iron(3+);trisulfate;hydrate
SMILES O.[Fe+3].[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 500 g
Linear Formula Fe2(SO4)3 · xH2O
UN Number 3260
Formula Weight 399.88g/mol (anhydrous basis)
Grade puriss. p.a.
Concentration or Composition (by Analyte or Components) 21 to 23% Fe basis
Packaging Poly bottle
Chemical Name or Material Iron(Iii) Sulfate Hydrate
Hazard Statement P280-P301 + P312 + P330-P305 + P351 + P338 + P310
Precautionary Statement H290-H302-H315-H318
RTECSNumber NO8520000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.