Learn More
Strem, An Ascensus Company CAS# 163169-29-7. 100MG. (S,S)-[2-(4'-i-Propyloxazolin-2'-yl)ferrocenyldiphenylphosphine, min. 97%. MFCD15144869
SDP
Supplier:  Strem, An Ascensus Company 264010100MG
Encompass
CAS# 163169-29-7. 100MG. (S,S)-[2-(4'-i-Propyloxazolin-2'-yl)ferrocenyldiphenylphosphine, min. 97%. MFCD15144869. Molecular Formula: C28H28FeNOP. Molecular Weight: 481.35. Color/Form: orange solid. Stability: air sensitive. Strem# 26-4010. www.strem.com/catalog/v3/26-4010/
Catalog No. 50-196-3573