Learn More
Strem, An Ascensus Company CAS# 541540-70-9. 500MG. (R,R)-[2-(4'-i-Propyloxazolin-2'-yl)ferrocenyl]diphenylphosphine, min. 97%. MFCD17018796
SDP
Catalog No. 501963530
Encompass
CAS# 541540-70-9. 500MG. (R,R)-[2-(4'-i-Propyloxazolin-2'-yl)ferrocenyl]diphenylphosphine, min. 97%. MFCD17018796. Molecular Formula: C28H28FeNOP. Molecular Weight: 481.35. Color/Form: orange solid. Stability: air sensitive. Strem# 26-4011. www.strem.com/catalog/v3/26-4011/
Catalog No. 50-196-3530 Supplier Strem, An Ascensus Company Supplier No. 264011500MG
CAS# 541540-70-9. 500MG. (R,R)-[2-(4'-i-Propyloxazolin-2'-yl)ferrocenyl]diphenylphosphine, min. 97%. MFCD17018796. Molecular Formula: C28H28FeNOP. Molecular Weight: 481.35. Color/Form: orange solid. Stability: air sensitive. Strem# 26-4011. www.strem.com/catalog/v3/26-4011/