Learn More
StatLab Iron Liver Human Control Slides
SDP
Catalog No. 21033341
Change view
Click to view available options
Quantity:
25 Pk.
Catalog No. Quantity
21-033-341 25 Pk.
1 options
Catalog No. 21-033-341 Supplier StatLab Supplier No. CSIRONL/25
Add to Cart
Add to Cart
Use iron liver control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 25 Unstained Slides per pack
Specifications
Type Control Slide
Quantity 25 Pk.