Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron nickel oxide

Catalog No. 637149100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-714-9100G 100 g
63-714-925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-714-9100G Supplier Sigma Aldrich Supplier No. 637149100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron nickel oxide (Fe2O3/NiO) nanopowder is a class of nanomaterial with properties such as superior magnetic, electrical and selective absorption. It can be used as an additive in the formation of membranes for water treatment.Fe2O3/NiO can be used for applications such as:fabrication of magnetic nanofiber for data storage and transferelectrocatalysts for the splitting of alkaline waterformation of composite nanofibers for efficient water oxidation electrocatalysis

Specifications

CAS 12168-54-6
Quantity 100 g
Molecular Formula Fe2NiO4
Linear Formula Fe2NiO4
MDL Number MFCD00016254
Synonym Diiron nickel tetraoxide
Molecular Weight (g/mol) 234.38
Percent Purity ≥98% trace metals basis
Density 5.368 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.