Learn More
Strem, An Ascensus Company CAS# 15321-51-4. 25g. Iron nonacarbonyl, 99%. MFCD00151465
SDP
Catalog No. 5090111112
Encompass
CAS# 15321-51-4. 25g. Iron nonacarbonyl, 99%. MFCD00151465. Molecular Weight: 363.79. Molecular Formula: Fe2(CO)9. Color/form: yellow to orange xtl. Strem# 26-2640. http://www.strem.com/catalog/v/26-2640/
Catalog No. 50-901-11112 Supplier Strem, An Ascensus Company Supplier No. 26264025G
CAS# 15321-51-4. 25g. Iron nonacarbonyl, 99%. MFCD00151465. Molecular Weight: 363.79. Molecular Formula: Fe2(CO)9. Color/form: yellow to orange xtl. Strem# 26-2640. http://www.strem.com/catalog/v/26-2640/