Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron oxide(II,III), magnetic nanoparticles solution

Catalog No. 7003125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-031-25ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-031-25ML Supplier Sigma Aldrich Supplier No. 7003125ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron oxide(II,III), magnetic nanoparticles solution (Fe3O4) is an organic solvent dispersed nanoparticle solution, which can be synthesized by the thermal decomposition of iron acetylacetonate in 2-pyrrolidone. It is majorly used in biomedical applications due to its magnetic properties, particle size, and surface properties.It contains <1.0% oleic acid stabilizing ligands.Fe3O4 can be used in the targeting and imaging of tumor cells for cancer therapy-based applications. It may also be used in the separation of tumor cells from fresh whole blood. It can also be used as an effective material in drug delivery systems and magnetic resonance imaging (MRI).

Specifications

CAS 1317-61-9
Quantity 5 mL
Molecular Formula Fe3O4
Linear Formula Fe3O4
MDL Number MFCD00011010
Synonym Magnetic iron oxide nanocrystals; Magnetite; Superparamagnetic iron oxide nanoparticles
Molecular Weight (g/mol) 231.53
Density 0.865 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.