Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iron oxide(II,III), magnetic nanoparticles solution

Catalog No. 7253315ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-533-15ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 72-533-15ML Supplier Sigma Aldrich Supplier No. 7253315ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iron oxide(II,III), magnetic nanoparticles solution is a chemically stable dispersed solution with a large surface to volume ratio. Its properties include good biocompatibility, and magnetic susceptibility (-t).Fe3O4 magnetic nanoparticles solution can be used in a variety of biomedical applications such as drug delivery. It can also be coated for immobilization of surface atoms for chelation of metal ions.

Specifications

CAS 1317-61-9
Quantity 5 mL
Molecular Formula Fe3O4
Linear Formula Fe3O4
MDL Number MFCD00011010
Synonym Magnetic iron oxide nanocrystals; Magnetite; Superparamagnetic iron oxide nanoparticles
Molecular Weight (g/mol) 231.53
Density 1.00 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.