Learn More
Sigma Aldrich Iron oxide (II,III), nanopowder
Catalog No. 9000341G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
Catalog No. Quantity
90-003-41G 1 g
1 options
Catalog No. 90-003-41G Supplier Sigma Aldrich Supplier No. 9000341G
Only null left
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Iron oxide nanoparticles exhibit biocompatibility and non-toxicity and are used as magnetic resonance imaging contrast agent, gene carrier for gene therapy, therapeutic agent for hyperthermia-cased cancer treatment, magnetic sensing probe for in-vivo diagnostics and target specific drug delivery.
Specifications
Quantity 1 g