Learn More
CHEMSCIENCE ChemScience Iron Oxide, Iron Ore, 99% Pure, 1 ea
Catalog No. NC3917263
Encompass_Preferred
This iron oxide product is a chemical grade iron ore intended for use in the analysis of elemental iron content.
  • Supplied as iron oxide
  • Validated for fe analysis
  • Manufactured in Canada
Catalog No. NC3917263 Supplier CHEMSCIENCE Supplier No. SI801300
This iron oxide product is a chemical grade iron ore intended for use in the analysis of elemental iron content.
  • Supplied as iron oxide
  • Validated for fe analysis
  • Manufactured in Canada