Learn More
Newcomer Supply Iron, Gomori Prussian Blue Stain Kit, 500 ml kit
SDP
Catalog No. NC2102277
Encompass
Newcomer Supply Iron, Gomori Prussian Blue Stain Kit procedure is used to detect loosely bound ferric iron in tissue sections. This histochemical reaction is sensitive enough to demonstrate minute amounts of iron deposits in blood cells, bone marrow and spleen.
This item is not returnable. View return policy
Catalog No. NC2102277 Supplier Newcomer Supply Supplier No. 9136B
Newcomer Supply Iron, Gomori Prussian Blue Stain Kit procedure is used to detect loosely bound ferric iron in tissue sections. This histochemical reaction is sensitive enough to demonstrate minute amounts of iron deposits in blood cells, bone marrow and spleen.