Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical™

Catalog No. SI124100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SI124-100 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SI124-100

missing translation for 'validMainframeMsg'

For atomic absorption. (1mL = 1mg Fe) Solute: Ferric Nitrate Solvent: Dilute Nitric Acid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-61-8 , 7697-37-2 , 7732-18-5
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Poly Bottle
Vapor Pressure 14mmHg
Physical Form Liquid
Concentration 990 to 1010 ppm
pH 1
Melting Point 0°C
Boiling Point 100°C
Quantity 100 mL
Grade Certified
Concentration or Composition (by Analyte or Components) 1000ppm ±1%
Chemical Name or Material Iron, Reference Standard Solution, 1000ppm ±1%
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.