Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Iron Refill Kit, Phenanthroline
SDP

Catalog No. 13299541
Change view
Click to view available options
Quantity:
30/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-541 30/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-541 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R6201
Add to Cart
Add to Cart

Refill kit for iron visual test kits measuring total and ferrous iron. Contains 30 self-filling ampoules packaged securely in trays and a box.

  • Ferrous iron reacts with 1,10-phenanthroline to form an orange-colored complex in direct proportion to the ferrous iron concentration.
  • Soluble iron consists primarily of ferrous iron but can also include a small portion of ferric iron. The soluble iron reagent contains hydroxylamine hydrochloride which converts any dissolved ferric iron to ferrous. The ferrous iron reagent does not contain hydroxylamine hydrochloride and will not measure soluble ferric iron. Total iron (ferric + ferrous) is determined by adding a solution of thioglycolic acid and ammonia (A-6000 Activator Solution) to the sample. This solution dissolves most forms of particulate iron and converts ferric iron to ferrous. Certain forms of very insoluble iron (magnetite, ferrite, etc.) require a digestion prior to analysis. Results are expressed as ppm (mg/L) Fe.

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-6210
No. of Tests 30
Quantity 30/Box
Type Iron Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.