Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Stain (Prussian Blue Iron) Kit, G-Biosciences
SCI_ED

Catalog No. S001125
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S001125 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S001125 Supplier G-Biosciences Supplier No. 7862235
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iron Stain is used in the histologic staining of iron in tissue or blood and bone marrow films. It is based on the well known Prussian blue reaction in which ionic iron reacts with acid ferrocyanide producing a blue color.

Each kit includes:

  • Potassium Ferrocyanide Solution, 4% w/v (1x 250 mL)
  • Hydrochloric Acid Solution, 1.2 mmol (1x 250 mL).
  • Pararosaniline hydrochloride, 1% w/v, (1x 30 mL).

Specifications

Quantity 1 Kit
Product Type Iron Stain Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.