Learn More
SPEX CERTIPREP LLC 3P Iron at 10,000 ug/mL in 10% HNO3, 100 mL
Catalog No. NC0911083
Encompass_Preferred
(HPS by Zeptometrix) Iron (Fe) ICP Single-Component Standard at 10,000 ug/mL in 10% HNO3.
Catalog No. NC0911083 Supplier SPEX CERTIPREP LLC 3P Supplier No. 10M261100
(HPS by Zeptometrix) Iron (Fe) ICP Single-Component Standard at 10,000 ug/mL in 10% HNO3.