Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iron Standard Stock Solution EP (200ppm) EP (200 ppm Fe), Reagents

Catalog No. 01334952
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-952 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-952 Supplier Reagents Holdings Llc Supplier No. CI002600120C
Add to Cart
Add to Cart

Iron Standard Stock Solution EP (200ppm) EP (200 ppm Fe), Reagent, use for European Pharmacopeia. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Iron Standard Stock Solution EP (200ppm) EP (200 ppm Fe)
Packaging Glass Bottle
CAS 7732-18-5, 7664-93-9
Specific Gravity 1.01
Physical Form Liquid
Color Pale Yellow/Green
pH 1.3
Boiling Point 100°C
For Use With (Equipment) Water Analyzer
Solubility Completely miscible
Quantity 120 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.