Learn More
Apexbio Technology LLC Iron sucrose 8047-67-4 100mg
SDP
Catalog No. 5000089679
Encompass_Preferred
Iron sucrose (CAS 8047-67-4) is a complex of polynuclear iron(III)-hydroxide with sucrose used as an intravenous iron supplement It delivers bioavailable iron which is incorporated into ferritin and hemoglobin following endocytosis and intracellular processing Unlike iron dextran iron sucrose is associated with a lower incidence of hypersensitivity reactions and is frequently used to manage iron deficiency anemia in chronic kidney disease both in patients undergoing dialysis and those who are not It is widely utilized in preclinical and clinical research to study iron metabolism erythropoiesis and anemia associated with renal disorders
Catalog No. 50-000-89679 Supplier Apexbio Technology LLC Supplier No. C6458100
Iron sucrose (CAS 8047-67-4) is a complex of polynuclear iron(III)-hydroxide with sucrose used as an intravenous iron supplement It delivers bioavailable iron which is incorporated into ferritin and hemoglobin following endocytosis and intracellular processing Unlike iron dextran iron sucrose is associated with a lower incidence of hypersensitivity reactions and is frequently used to manage iron deficiency anemia in chronic kidney disease both in patients undergoing dialysis and those who are not It is widely utilized in preclinical and clinical research to study iron metabolism erythropoiesis and anemia associated with renal disorders