Learn More
NEOGEN Iron Sulphite Agar, 500 g
SDP
Catalog No. 502015771
Encompass_Preferred
Iron Sulphite Agar is a medium for the detection of thermophilic anaerobic organisms causing sulphide spoilage in food, and is not intended for use in the diagnosis of disease or other conditions in humans.This formulation is a modification of Cameron Sulphite Agar, which was developed by the National Canners Association of America (now the Grocery Manufacturers Association). Iron Sulphite Agar has a reduced concentration of sodium sulphite to allow improved detection of some strains of Clostridium sporogenes. Beerens, and later Mossel, demonstrated that some strains of C. sporogenes would not tolerate sodium sulphite levels of 0.1%. Mossel further observed that reducing sulphite content to 0.05% improved detection of these strains.For more information visit https://www.neogen.com/solutions/Microbiology/iron-sulphite-agar
Catalog No. 50-201-5771 Supplier NEOGEN Supplier No. 700004654
Iron Sulphite Agar is a medium for the detection of thermophilic anaerobic organisms causing sulphide spoilage in food, and is not intended for use in the diagnosis of disease or other conditions in humans.This formulation is a modification of Cameron Sulphite Agar, which was developed by the National Canners Association of America (now the Grocery Manufacturers Association). Iron Sulphite Agar has a reduced concentration of sodium sulphite to allow improved detection of some strains of Clostridium sporogenes. Beerens, and later Mossel, demonstrated that some strains of C. sporogenes would not tolerate sodium sulphite levels of 0.1%. Mossel further observed that reducing sulphite content to 0.05% improved detection of these strains.For more information visit https://www.neogen.com/solutions/Microbiology/iron-sulphite-agar