Learn More
HIMEDIA Iron Sulphite Agar Modified, M1852I, Microbiology Culture Media, Dehydrated Culture Media, Agar, Sulphite-Reducing Bacteria, ISO 15213-1 and ISO 11133, Powder, 500 g
Catalog No. NC4013706
Encompass_Preferred
Iron Sulphite Agar Modified is recommended for enumerating sulphite-reducing bacteria, including clostridia, growing under anaerobic conditions, with composition and performance aligned to ISO 15213-1 (2023) and ISO 11133. Peptone, soya digest and yeast extract nourish the organisms while sulphite and an iron source act as the indicator system. Reduction of sulphite produces iron sulphide and characteristic black colonies that are easily counted. Widely used in food and water microbiology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013706 Supplier HIMEDIA Supplier No. M1852I500G
Iron Sulphite Agar Modified is recommended for enumerating sulphite-reducing bacteria, including clostridia, growing under anaerobic conditions, with composition and performance aligned to ISO 15213-1 (2023) and ISO 11133. Peptone, soya digest and yeast extract nourish the organisms while sulphite and an iron source act as the indicator system. Reduction of sulphite produces iron sulphide and characteristic black colonies that are easily counted. Widely used in food and water microbiology. Dehydrated powder, store in a cool dry place. Packaged as 500 g.