Learn More
Hach Company Iron TNTplus Vial Test (0.2-6.0 mg/L Fe), 25 Tests
Supplier:  Hach Company TNT858
Encompass
Iron TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.For determination of Iron by the Phenanthroline method. Add Metals Prep Set (TNT890) for total iron determination (requires digestion).
Catalog No. 50-199-8145