Learn More
Hach Company Iron trace Vial Test, ULR (0.01 - 1 mg/L Fe), 20 Tests
Supplier:  Hach Company HPT251
Encompass
Iron HPT chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime. For determination of very low Iron concentrations, e.g. in industrial or municipal water samples by the 1.10-Phenanthroline Method. Convenient handling using 50 mm rectangular vials. Optional digestion with TNT890 to evaluate total iron. 20 ULR Iron vials per box.
Catalog No. 50-199-7624