Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Iron Water Quality Visual Test Kit, 250 to 2500 ppm
SDP

Catalog No. 13299675
Change view
Click to view available options
No. of Tests:
30
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
13-299-675 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-299-675 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K6020B
Add to Cart
Add to Cart

Iron visual test kit featuring self-filling ampoules with pre-measured reagent for a single test. Compare results to color standards to determine concentration.

  • The K-6020B Iron (total & soluble) HR CHEMets™ Test Kit (250-2500 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): HR CHEMets™ Refill, Flat Comparator, 25 mL sample cup, pipettor tips, 100 uL fixed volume pipet, iron activator solution, and instructions.

Specifications

Analytes Iron
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 250 to 2500 ppm
Type Water Quality Visual Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.