Learn More
MSC PIP Ironcat Welding Gloves, Split Cowhide, Aramid Lined, 14 in., 72/Case
Catalog No. 5000688942
Encompass
These premium grade welding gloves are constructed from split cowhide leather, offering a balance of comfort, durability, and breathability. Designed for professional protection, they feature a para-aramid blended lining that provides 360 degree cut and heat resistance extending through the cuff. The gloves are reinforced for demanding work environments and include high-performance arc protection.
  • Features a 63 cal/cm2 ARC rating (ATPV) for thermal arc flash protection
  • Lined with para-aramid for ANSI A4 cut resistance and level 3 puncture protection
  • Reinforced palm and thumb strap enhance durability at high-wear contact points
  • Stitched with Kevlar for superior strength and burn resistance
  • Wing thumb design improves comfort during open-handed tasks
  • Extended 14 in. gauntlet cuff provides additional forearm coverage
Catalog No. 50-006-88942 Supplier MSC Supplier No. 21004940
Add to Cart
Add to Cart
These premium grade welding gloves are constructed from split cowhide leather, offering a balance of comfort, durability, and breathability. Designed for professional protection, they feature a para-aramid blended lining that provides 360 degree cut and heat resistance extending through the cuff. The gloves are reinforced for demanding work environments and include high-performance arc protection.
  • Features a 63 cal/cm2 ARC rating (ATPV) for thermal arc flash protection
  • Lined with para-aramid for ANSI A4 cut resistance and level 3 puncture protection
  • Reinforced palm and thumb strap enhance durability at high-wear contact points
  • Stitched with Kevlar for superior strength and burn resistance
  • Wing thumb design improves comfort during open-handed tasks
  • Extended 14 in. gauntlet cuff provides additional forearm coverage