Learn More
MSC PIP Ironcat 8051 FR Cotton Cape Sleeve, Proban Cotton, Leather Reinforced, 7XL, 1 ea
Catalog No. 5000688941
Encompass
This flame-resistant cape sleeve with apron provides lightweight protection for industrial applications such as steel mills. Constructed with premium cow split leather sleeves and a Proban treated cotton woven fabric, the garment ensures long-lasting flame resistance. It is compliant with NFPA 2112, NFPA 70E PPE2, ASTM F1959, and ASTM F1506 standards.
  • Inherently flame-resistant cotton fabric
  • Leather reinforced snaps to prevent flashback
  • Snap at wrist for a secure fit
  • Lay-down or pop-up collar feature
  • Flame resistance lasts for up to 100 home washings or 50 commercial washings
  • Compliant with NFPA 2112, NFPA 70E PPE2, ASTM F1959 and ASTM F1506
Catalog No. 50-006-88941 Supplier MSC Supplier No. 21004015
This flame-resistant cape sleeve with apron provides lightweight protection for industrial applications such as steel mills. Constructed with premium cow split leather sleeves and a Proban treated cotton woven fabric, the garment ensures long-lasting flame resistance. It is compliant with NFPA 2112, NFPA 70E PPE2, ASTM F1959, and ASTM F1506 standards.
  • Inherently flame-resistant cotton fabric
  • Leather reinforced snaps to prevent flashback
  • Snap at wrist for a secure fit
  • Lay-down or pop-up collar feature
  • Flame resistance lasts for up to 100 home washings or 50 commercial washings
  • Compliant with NFPA 2112, NFPA 70E PPE2, ASTM F1959 and ASTM F1506