Learn More
MSC IRONCAT TWL CTN FR JCKT
Catalog No. 5000686608
Encompass
The Ironcat FR Treated 100% Cotton Sateen Jacket is designed for robust protection in demanding industrial environments, including steel mills, shipyards, manufacturing, automotive, gas welding, and torch work.
  • FR treated 9 oz. 100% cotton sateen fabric provides protection from sparks.
  • Anodized snaps are non-conductive and corrosion-resistant, essential for safety and performance.
  • A lower interior pocket offers protected storage for small personal items or tools.
  • Includes a black pop-up welder's collar with snap closure.
  • A scribe-soapstone pocket is located on the left sleeve for easy access.
  • Meets standards: ISO 11611, ISO 11612, ASTM D6413.
Catalog No. 50-006-86608 Supplier MSC Supplier No. 47749932
The Ironcat FR Treated 100% Cotton Sateen Jacket is designed for robust protection in demanding industrial environments, including steel mills, shipyards, manufacturing, automotive, gas welding, and torch work.
  • FR treated 9 oz. 100% cotton sateen fabric provides protection from sparks.
  • Anodized snaps are non-conductive and corrosion-resistant, essential for safety and performance.
  • A lower interior pocket offers protected storage for small personal items or tools.
  • Includes a black pop-up welder's collar with snap closure.
  • A scribe-soapstone pocket is located on the left sleeve for easy access.
  • Meets standards: ISO 11611, ISO 11612, ASTM D6413.