Learn More
Ironclad Performance Wear Ironclad Box Handler Glove, 12 pr per box, size Small, Black, EN388
Catalog No. 501610019
Encompass_Preferred
Gloves, Ironclad, Box Handler Glove, Extreme Grip, Breathable nylon back of hand, Silicone printed microsuede palm, Neoprene Knuckle, Thumb nail guard, Hybrid safety cuff. Specifically designed for parcel delivering, automotive repair, glass, warehouse application and drywall handling. 12 pr per box.
This item is not returnable. View return policy
Catalog No. 50-161-0019 Supplier Ironclad Performance Wear Supplier No. BHG02S
Gloves, Ironclad, Box Handler Glove, Extreme Grip, Breathable nylon back of hand, Silicone printed microsuede palm, Neoprene Knuckle, Thumb nail guard, Hybrid safety cuff. Specifically designed for parcel delivering, automotive repair, glass, warehouse application and drywall handling. 12 pr per box.