Learn More
Ironclad Performance Wear Ironclad Cold Condition Glove, 12 pr per box, size Small Black, EN388
Catalog No. 501610024
Encompass_Preferred
Gloves, Ironclad, Cold Condition Glove, 30 degree F water-resistant, Duraclad reinforced palm, Wind and Water resistant construction, Microfleece liner, TPR knuckle impace protection, Reflective back of hand print, Neoprene compression cuff. Ideal for any task where precision is desired in cold weather, cold storage, equipment operation. 12 pr per box
This item is not returnable. View return policy
Catalog No. 50-161-0024 Supplier Ironclad Performance Wear Supplier No. CCG202S
Gloves, Ironclad, Cold Condition Glove, 30 degree F water-resistant, Duraclad reinforced palm, Wind and Water resistant construction, Microfleece liner, TPR knuckle impace protection, Reflective back of hand print, Neoprene compression cuff. Ideal for any task where precision is desired in cold weather, cold storage, equipment operation. 12 pr per box