These gloves are designed for versatility and superior jobsite protection. They feature a synthetic suede palm for a natural feel and wear resistance, and an impact-resistant back with flexible rubber fingers, thumb, and knuckle guards. The palm is reinforced and padded for hard use, and a cut-resistant liner protects both the palm and back of the hand.
- Synthetic suede palm for natural feel and wear resistance
- Impact-resistant back via flexible rubber fingers, thumb, and knuckle guard
- Reinforced and padded palm for hard use
- Cut-resistant liner protects palm and back of hand
- Touchscreen compatible
- Velcro closure for easy entry and secure fit
- Terry cloth sweat wipe thumb
- Complete 360° Cut A6 HPPE blend liner
- TPR impact protection fingers, thumb, and knuckle
- Thumb saddle reinforcement